Learn More
Fisherbrand™ Reverse Osmosis Membrane with Housing
Deionizes tap water and filters out particles for laboratory procedures. Fisher Scientific™ Reverse Osmosis Membrane with Housing is for use with the Accu500 Ultrapure Water System.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.40000kg
Descrizione
- For use with Accu500.
- Two sizes: 10L / 20L or 15L / 30L
- Removes particles, ions, and organics from tap water
Specifica
Accu500 15/30L | |
Reverse Osmosis Membrane | |
Housing |