Learn More
Hirschmann™ ringcaps™ for Reflotron™ Disposable Capillary Pipettes
Capillary pipettes with capacity line, for the Roche Reflotron™ system.
Marca: Hirschmann™ 9550532
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- With ringmark
- Conformity certified
- Manufactured from durable DURAN™ glass
- Lithium-heparinized
- 32 μL
- Capillary for Reflotron™ system
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifica
Disposable capillary pipettes with ringmark at 32 μL, Lithium heparinized | |
ringcaps™ | |
Reflotron |
Borosilicate Glass | |
Single-use |