missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17563-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
RIP3 is a novel member of RIP Ser/Thr kinase family. It is a potent inducer of apoptosis and is an important component of TNFR1 signaling complex. RIP3 consists of an N-terminal RIP-like-Kinase domain and a unique, non-homologous C-terminus responsible for caspase activation and apoptosis induction. RIP3 is expressed in multiple tissues including hematopoietic cells and plays a functional role in the regulation of apoptosis and NF-kB signaling. It is recruited to the TNFR1 signaling complex in a RIP-dependant manner where it induces apoptosis by activating caspases and/or inhibiting TNFR1-induced NF-kB activation. RIP3 negatively regulates the TLR3-Trif-mediated NF-kB signaling pathway by competing with the binding of Trif to RIP1. The human RIP3 gene is localized in the chromosomal region 14q11.2.Specifica
RIP3 | |
Polyclonal | |
Unconjugated | |
RIPK3 | |
Receptor interacting protein 3, RIP 3, RIP like protein kinase 3, RIP3, RIPK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11035, 56532 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QZL0, Q9Y572 | |
Ripk3 | |
RIP3 Fusion Protein Ag11759 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |