Learn More
Corning™ rLaminin-521 (Human)
Recombinant heterotrimer protein composed of α5, β2, and γ1 chains
Marca: Corning™ 354223
Dettagli aggiuntivi : Peso : 0.13900kg
Descrizione
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning rLaminin-521 is the next generation pluripotent stem cell (PSC) surface that enables researchers to streamline culture workflow, lower contamination risk, and scale-up efficiently. Corning has partnered with BioLamina for the supply of recombinant human Laminin 521 (rLaminin-521), a biocompatible, full length Laminin that enables feeder-free, clump-free, single-cell passaging of PSCs without ROCK-inhibitor.
Specifica
rLaminin-521, Human | |
5 mg | |
Human | |
Recombinant |