missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF156 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11285-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.Specifica
RNF156 | |
Polyclonal | |
Unconjugated | |
MGRN1 | |
KIAA0544, mahogunin, ring finger 1, MGRN1, RING finger protein 156, RNF156 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17237, 23295, 302938 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60291, Q5XIQ4, Q9D074 | |
Mgrn1 | |
RNF156 Fusion Protein Ag1808 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |