Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Marca: SVL™ RF35
1216.00 EUR valido fino al 2024-12-31
Usa il codice promo "23259" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.39000kg
Descrizione
- Offers a grease free spherical joint
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200°C
- System comprises male ball joint with PTFE covered O-ring which fits into a female cup joint
- Joint can be retained together with spring loaded metal clipComponent parts RM, RF, RO and RC should be ordered separately
Specifica
Female joint, PTFE covered O-ring, spring loaded Metal clip | |
Glass, PTFE | |
Spherical joint | |
35 |