Learn More
RPE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12168-2-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The function of this protein remains unknown.Specifica
| RPE | |
| Polyclonal | |
| Unconjugated | |
| RPE | |
| ribulose 5 phosphate 3 epimerase, Ribulose phosphate 3 epimerase, RPE, RPE2 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 501157, 6120, 66646 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VEE0, Q96AT9 | |
| RPE | |
| RPE Fusion Protein Ag2810 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |