missing translation for 'onlineSavingsMsg'
Learn More

RPL8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 16981-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896784

  • 129.00€ / 20µL

Spedizione stimata: 23-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L2P family of ribosomal proteins. It is located in the cytoplasm. In rat, the protein associates with the 5.8S rRNA, very likely participates in the binding of aminoacyl-tRNA, and is a constituent of the elongation factor 2-binding site at the ribosomal subunit interface. Alternatively spliced transcript variants encoding the same protein exist. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifica

Specifica

RPL8
Polyclonal
Unconjugated
RPL8
60S ribosomal protein L8, ribosomal protein L8, RPL8
Rabbit
Antigen Affinity Chromatography
RUO
26961, 26962, 6132
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62917, P62918, P62919
RPL8
RPL8 Fusion Protein Ag9957
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali