Learn More
RSBN1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554199
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KRPKMYSKSI QTICSGLLTD VEDQAAKGIL NDNIKDYVGK NLDTKNYDSK IPENSEFPFV SLKEPRVQNN LKRLDTLEFK QLIHIEHQPN GGASVIHAYS NELSHLS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.
RSBN1L Polyclonal antibody specifically detects RSBN1L in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
RSBN1L | |
Polyclonal | |
Unconjugated | |
RSBN1L | |
8430412F05Rik; AI447441; C330002G24Rik; Lysine-specific demethylase RSBN1L; RGD1308748; round spermatid basic protein 1 like; round spermatid basic protein 1-like; round spermatid basic protein 1-like protein; Rsbn1l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
222194, 242860, 311987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PCB5 | |
RSBN1L | |
Recombinant protein corresponding to Human RSBN1L | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only