Learn More
3M™ S-Series Hood with Inner Shroud, Premium Head Suspension
S-Series hood with sealed seams features a double shroud design for its respiratory seal.
1090.00€
Specifica
Assigned Protection Factor (APF) | 1000 |
---|---|
Materiale | Zytron 200 |
Da utilizzare con (applicazione) | Assembly, facility maintenance, machine operation, painting, sanding |
Da utilizzare con (apparecchiatura) | GVP, Breathe-Easy™, V Series, T-300, T-600, T-800 |
Tipo di sospensione | Premium Resusable Suspension |
Codice del prodotto | Marca | Product Type | Include | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Product Type | Include | Prezzo | Quantità e disponibilità | ||||
13481177
|
3M™
7100079873 |
Hood with Neck Collar | Hood with neck collar only |
1090.00€
pezzi 5 |
Spedizione stimata: 11-07-2024
Eseguire il login per visualizzare lo stock disponibile |
||||
Descrizione
Premium suspension hoods and head covers
- Head covers provide head and face coverage; hoods cover head, face, and neck
- Fully assembled and ready to use
- Suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or situations where there is less soiling of the outer fabric
- Integrated models feature wide face seal with soft elastic for improved comfort and reduced noise
Premium Reusable Suspensions with Hoods
- Ideal for high disposal rate environments
- Soiled hood is replaced; suspension and air ducting are reused
- Rigid ducting for consistent air path
- Externally adjustable airflow control allows wearer to modify airflow distribution
- Suspension features independent adjustments for the crown and head circumference; allows custom fit for wide range of head sizes
- 3M Catalog# S-805-5, S-807-5, S-855, S-857
Specifica
1000 | |
Assembly, facility maintenance, machine operation, painting, sanding | |
Premium Resusable Suspension |
Zytron 200 | |
GVP, Breathe-Easy™, V Series, T-300, T-600, T-800 | |
Acetate |