Learn More
Thermo Scientific™ Samco™ Tube Closures
Depend on the reliable performance of these tube closures, available as flanged caps, two-position snap caps and universal caps.
Marca: Thermo Scientific™ M995133
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
Depend on the reliable performance of Thermo Scientific™ Samco™ Tube Closures. Choose from two-position snap caps and universal caps. Our processes are monitored with quality checks which provide continued lot-to-lot uniformity and reliable performance.
Specifica
Universal Plug Cap | |
Yellow | |
Non-sterile |
Non-sterile universal plug cap for 12-16mm tubes | |
10000 |