Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD (E484K), His Tag
MALS verified
Marca: ACROBiosystems SRD-C52H3-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
QHD43416.1 (E484K) | |
1 mg | |
Arg 319 - Lys 537 | |
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
26.5 kDa | |
SARS-CoV-2 S protein RBD (E484K), His Tag (SRD-C52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (E484K)). E484K mutaion was identified in the SARS-CoV-2 variant (known as 20 C/501 Y.V2 or B0.1.351 lineage) which emerged in South Africa. | |
-20°C | |
S protein RBD | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |