Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein NTD, His Tag
Marca:  ACROBiosystems S1D-C52H6-1mg
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17106671
  • dietro preventivo / 1mg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Specifica
Specifica
QHD43416.1
34.9 kDa
1 mg
Ser 13 - Leu 303
S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19
Unconjugated
PBS,pH7.3
HEK293
SARS-CoV-2
Powder
>90% as determined by SDS-PAGE
SARS-CoV-2 S1 protein NTD, His Tag (S1D-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1).
-20°C
S1 protein NTD
His Tag
Less than 1.0 EU / µg by the LAL method.
Viral Proteins