missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (75G5a), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537453
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.Specifica
SARS-CoV-2 Nucleocapsid | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
P0DTC9 | |
N | |
SARS-CoV-2 nucleoprotein (NP) specific peptide. | |
20 μL | |
Primary | |
Virus | |
Antibody | |
IgG κ |
ELISA | |
75G5a | |
Unconjugated | |
N | |
COVID19 Nucleocapsid, COVID-19 nucleocapsid, COVID19 Spike nucleocapsid, SARS-CoV-2 N | |
Rabbit | |
Protein A | |
RUO | |
43740575 | |
4° C | |
Liquid |