missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (75G5a), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537205
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
SARS-CoV-2 Nucleocapsid Recombinant Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in Virus samples. It is validated for ELISA.Specifica
SARS-CoV-2 Nucleocapsid | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
P0DTC9 | |
N | |
SARS-CoV-2 nucleoprotein (NP) specific peptide. | |
200 μL | |
Primary | |
Virus | |
Antibody | |
IgG κ |
ELISA | |
75G5a | |
Unconjugated | |
N | |
COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N | |
Rabbit | |
Protein A | |
RUO | |
43740575 | |
4° C | |
Liquid |