missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 ORF8 (aa16-121), His Tag Recombinant Protein
E. coli Recombinant Protein
Marca: Invitrogen™ RP87666
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
ORF8 is a SARS-CoV accessory gene. The exact role of ORF8 in pathogenesis in unknown; however, among the unique accessory ORFs, ORF8 is most hypervariable.Specifica
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
1 mg | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
2019-nCoV orf8; Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 orf8; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS-CoV orf8; SARS-CoV-2; SARS-CoV-2 orf8 | |
Unconjugated | |
Liquid |
Control, ELISA, Western Blot | |
SARS-CoV-2 ORF8 (aa16-121), His Tag | |
E. coli | |
RUO | |
Virus | |
Recombinant | |
>95% |