Learn More
ACROBiosystems SARS-CoV-2 S protein (HV69-70del, Y144del, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H), His Tag (MALS verified)
Marca:  ACROBiosystems SPN-C52H6-500ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17226550
  • dietro preventivo / 500µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 137.8 kDa. The protein migrates as 150-180 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
QHD43416.1
>90% as determined by SDS-PAGE.
SARS-CoV-2 S protein, His Tag (SPN-C52H6) is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 (Accession # QHD43416.1) and HV69-70 del, Y144del, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H mutations, which have become increasingly common in SARS-CoV-2 viruses from around the world. The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. HV69-70 del / Y144del / N501Y / A570D / D614G / P681H / T716I / S982A / D1118H mutations were identified in the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom.)
-20°C
S protein
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
SARS-CoV-2
137.8 kDa
500 µg
Val 16 - Pro 1213
Spike,S protein,Spike glycoprotein,S glycoprotein,COVID-19
Unconjugated
PBS
HEK293
Viral Proteins