Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (K417N, E484K, N501Y), Fc Tag
Marque: ACROBiosystems SPD-C5256-1mg
Informations supplémentaires : Peso : 0.00010kg
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 51.1 kDa.Spécification
51.1 kDa | |
SARS-CoV-2 S protein RBD (K417N,E484K,N501Y), Fc Tag (SPD-C5256) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417N, E484K, N501Y)). | |
-20°C | |
S protein RBD | |
SARS-CoV-2 | |
HEK293 | |
Viral Proteins |
1 mg | |
Arg 319 - Lys 537 | |
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 | |
Unconjugated | |
Fc Tag | |
Powder |