Learn More
ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, R246I, K417N, E484K, N501Y, D614G), His Tag
Marca:  ACROBiosystems S1N-C52Hm-100ug
Dettagli aggiuntivi : Peso : 0.00010kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17246470
  • dietro preventivo / 100µg
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Descrizione
Descrizione
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.8 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica
Specifica
QHD43416.1
>90% as determined by SDS-PAGE.
SARS-CoV-2 S1 protein, His Tag (S1N-C52Hm) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1). L18F/ D80A/ D215G/ R246I/ K417N/ E484K/ N501Y/ D614G mutations were identified in the SARS-CoV-2 variant (known as 20 C/501 Y.V2 or B0.1.351 lineage) which emerged in South Africa.
-20°C
S1 protein
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
SARS-CoV-2
76.8 kDa
100 µg
Val 16 - Arg 685
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
PBS, pH7.4
HEK293
Viral Proteins