Learn More
Sartorius Qualitative Grade 39/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Marca:  Sartorius FT-4-483-385
Dettagli aggiuntivi : Peso : 2.42000kg
Codice prodotto. 15953010
  • 660.00€ / pezzi 100
Spedizione stimata: 03-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.
Specifica
Specifica
Folded
39/N
0.65 mm
5 sec.
100/Pk.
385 mm