Learn More
SBSN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA564338
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: HQAGKEAEKL GQGVNHAADQ AGKEVEKLGQ GAHHAAGQAG KELQNAHNGV NQASKEANQL LNGNHQSGSS Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 76%.
SBSN is a protein coding gene.Specifica
SBSN | |
Polyclonal | |
Unconjugated | |
SBSN | |
HLAR698; SBSN; Suprabasin; UNQ698; UNQ698/PRO1343 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWP8 | |
SBSN | |
Recombinant protein corresponding to Human SBSN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only