Learn More
SI Analytics™ ScienceLine™ pH Electrode
SI Analytics™ ScienceLine™ pH Electrode is a metal and glass electrode with a Silamid™ reference system.
Marca: SI Analytics™ 285103607
Dettagli aggiuntivi : Peso : 0.12000kg
Descrizione
- Reference system: Silamid™
- Glass shaft
- Ag 6280
Specifica
Plug Head | |
pH Electrode | |
-5°C to +100°C | |
4 mm sensor |
Glass Shaft | |
Sphere | |
120 mm | |
12 mm |