Learn More
SCLT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA557717
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KTQAVELWQT VSQELDRLHK LYQEHMTEAQ IHVFESQKQK DQLFDFQQLT KQLHVTNENM EVTNQQFLKT VTEQSVIIE Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
Adapter protein that links SCN10A to clathrin. Regulates SCN10A channel activity, possibly by promoting channel internalization.Specifica
SCLT1 | |
Polyclonal | |
Unconjugated | |
SCLT1 | |
2610207F23Rik; 4931421F20Rik; CAP1A; CAP-1A; Sap1; SCLT1; Sodium channel and clathrin linker 1; sodium channel-associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96NL6 | |
SCLT1 | |
Recombinant protein corresponding to Human SCLT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only