Learn More
SDSL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552196
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: RHLVCSSGGN AGIAAAYAAR KLGIPATIVL PESTSLQVVQ RLQGEGAEVQ LTGKVWDEAN LRAQELAKRD GWENVPPFDH PLIWKGHASL VQELKAVLRT Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
SDSL has low serine dehydratase and threonine dehydratase activity.Specifica
SDSL | |
Polyclonal | |
Unconjugated | |
SDSL | |
4432411H13Rik; AI504310; cSDH; L-serine deaminase; L-serine dehydratase/L-threonine deaminase; L-threonine dehydratase; SDH; SDH 2; SDH1; Sds; SDSL; SDS-RS1; Serine dehydratase 2; serine dehydratase like; serine dehydratase related sequence 1; serine dehydratase-like; TDH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113675, 360816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96GA7 | |
SDSL | |
Recombinant protein corresponding to Human SDSL | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Research Use Only