missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Edmont™ Guanti in lattice talcati Semperguard™
Click to view available options
Size:
6.5
8.5
Descrizione
Ideal for use in laboratories with short-term chemical contact, light duty maintenance, painting, printing, pharmaceutical production and applications requiring protection and sensitivity.
- Resistant to many cleaning chemicals used in laboratories
- Flexible and comfortable for excellent dexterity
- Lightly flocked interior for better sensitivity
- Sandblasted finish to guarantee a secure grip in dry or wet conditions
- Extended-length, tear-resistant beaded cuff for extra protection and easy donning
- Fluted cuff design enables ventilation to the hand
Specifica
Certificazione/Conformità | EN420 (General requirements), EN388 (Mechanical Hazards) and EN374 (Chemical and Microorganisms Hazards) |
Classificazione | Manufactured under EC quality assurance system carried out by the BSI. |
Colore | Red |
Finitura esterna | Sand Blast |
Hand | Specific |
Latex Free | No |
Materiale | Rubber Latex |
Size | 8.5 |
Spessore (sistema metrico) | 0.70 mm |
Stile polso guanto | Beaded, Fluted |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'