missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SemperIt™ Semperguard™ Nitrile Xpert Protective Gloves
Click to view available options
2XL
Descrizione
Protects against chemical splashing and in the processing of fatty foods. Provides excellent chemical resistance to skin irritants.
- Manufactured from high quality soft blue nitrile film
- Textured outer surface for better grip especially in wet conditions
- Ambidextrous fit with rolled cuff
Specifica
Certificazione/Conformità | CE Class I, EN 420, EN 374, EN 455, PPE Category III |
Colore | Blue |
Monouso | Single-use |
Hand | Ambidextrous |
Latex Free | Yes |
Materiale | Nitrile |
Powdered | Powder Free |
Size | 2X-Large |
Spessore (sistema metrico) | 0.20 mm |
Stile polso guanto | Rolled |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'