missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SemperIt™ Semperguard™ Semperplus™ Nitrile Protective Gloves
Click to view available options
Medium (8)
Descrizione
- Extra length for higher safety (270-285 mm)
- Extra wall thickness
- Straight edge cuff style
- Green
Specifica
Certificazione/Conformità | CE, EN 420, EN 374, EN 388, PPE Category III |
Coating Material | Diamond pattern outside |
Colore | Green |
Monouso | Reusable |
Latex Free | Yes |
Materiale | Nitrile |
Size | Medium, 8 |
Spessore (sistema metrico) | 0.38 mm |
Stile polso guanto | Straight |
Rivestimento interno | Flock lined with Cotton |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'