missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SemperIt™ Semperguard™ SemperStar™ Vinyl Protective Gloves
Click to view available options
Large (9)
Specifica
Certificazione/Conformità | CE, EN 420, EN 374, EN 388, PPE Category III |
Colore | Violet |
Monouso | Reusable |
Finitura esterna | Structured |
Materiale | Vinyl |
Size | Large, 9 |
Spessore (sistema metrico) | 0.6 mm |
Da utilizzare con (applicazione) | Working with chemicals, work safety, printing houses, forestry, gardening, household |
Stile polso guanto | Waved |
Rivestimento interno | Flock lined with Cotton |
Vedi altri risultati |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'