Learn More
SERPINB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA530015
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Regulates the activity of the neutrophil proteases elastase, cathepsin G, proteinase-3, chymase, chymotrypsin, and kallikrein-3.Specifica
SERPINB1 | |
Polyclonal | |
Unconjugated | |
SERPINB1 | |
1190005M04Rik; AI325983; Anti elastase; anti-elastase; EGK_14416; EI; EIA; ELANH2; epididymis luminal protein 57; epididymis secretory protein Li 27; HEL57; HEL-S-27; L M/N MN PI2; LEI; leukocyte elastase inhibitor; Leukocyte elastase inhibitor A; M/NEI; MNEI; Monocyte/neutrophil elastase inhibitor; ovalbumin; peptidase inhibitor 2; PI2; PI-2; protease inhibitor 2 (anti-elastase), monocyte/neutrophil derived; serine (or cysteine) peptidase inhibitor, clade B, member 1a; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 1a; serine (or cysteine) proteinase inhibitor, clade B, member 1a; serine protease inhibitor EIA; serpin; Serpin B1; Serpin B1a; serpin family B member 1; serpin peptidase inhibitor, clade B (ovalbumin), member 1; Serpinb1; Serpinb1a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1992 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.67 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P30740 | |
SERPINB1 | |
Recombinant fragment corresponding to a region within amino acids 117 and 379 of Human M/NEI | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |