missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11772-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
May have the capability to bind to carbohydrates.Specifica
SFRS6 | |
Polyclonal | |
Unconjugated | |
SRSF6 | |
B52, Pre mRNA splicing factor SRP55, SFRS6, SRP55, SRSF6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362264, 6431, 67996 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
G3V6S8, Q13247, Q3TWW8 | |
Srsf6 | |
SFRS6 Fusion Protein Ag2412 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |