Learn More
Trajan™ Thermo Scientific Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Marca: Trajan™ 073452
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifica
SilTite Metal | |
GC Interface Connection | |
10 ferrules, 2 nuts | |
0.45 to 0.53 mm |