Learn More
Epredia™ Shandon™ Colormark™ Plus Slides
Improve cellular adhesion with positively charged Epredia™ Shandon™ ColorMark™ Plus Slides.
Marca: Epredia™ B9992010CS
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
Available in a variety of colors.
Specifica
Microscope Slide | |
Glass | |
Pink | |
144/Pk. |