Learn More
Trajan™ Shimadzu Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Marca: Trajan™ 073227
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifica
SGE; GC14 A, GC17 A, GC2010 and GC2014 Detector/Injectors | |
SilTite Metal | |
Ferrule |