Learn More
Trajan™ Shimadzu Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Marca: Trajan™ 0726564
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifica
QP5000/5050 Standard MS Interface | |
450°C | |
GC-MS | |
350°C | |
0.5 mm |
0.019 in. | |
662°F | |
15% Graphite, 85% Vespel | |
Ferrule |