Learn More
Anti-schiuma al silicio
Click to view available options
Quantità:
1 L
Imballaggio:
Polyethylene
Descrizione
Foam reduces processing speed and capacity in many laboratory applications. A small amount of silicon antifoam prevents the formation of foam and rapidly breaks down foam that is already present. A foam’s film is stabilized by its surface elasticity and viscosity. Silicones have low surface tension and interfacial properties. Their lack of foam-stabilization properties causes foam walls to become thin and collapse.
Specifica
Densità | 1.00g/mL |
Quantità | 1 L |
Imballaggio | Polyethylene |
Dimensione particelle | 1LT |
Volume dei pori | 1LT |