Learn More
Cytiva Whatman™ Replacement Silicone O-Ring for MD050 Series Pressure Filtration Manifolds
Replacement Silicone O-Ring for MD050 Series Pressure Filtration Manifolds.
Marca: Cytiva 10450009
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Product Type: O-Ring
- Model: MD050/0/31
- Material: Silicone
- For Use With (Application): Pressure Filtration
- For Use With (Equipment): MD050 Series Pressure Filtration Manifolds
Specifica
Replacement O-Ring; Model: MD050/0/31 | |
Pressure Filtration | |
MD050 Series Pressure Filtration Manifolds |
O-Ring | |
Silicone |