Learn More
Kartell™ K70 Silicon Hoses
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
Marca:  Kartell™ 0392900
Dettagli aggiuntivi : Peso : 0.25000kg
Codice prodotto. 10063311
  • 129.00€ / 10m
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
  • Flexible and transparent
  • FDA, USP, Class VI, A 50p60, EPC VI.1.3.2 approved
  • Can withstand temperatures of -60°C to 200°C
Specifica
Specifica
FDA, USP, Class VI, A 50p60 and EPC VI.1.3.2 Approved
10 mm
Silicone
10 m
Transparent
Max. Operating Temperature (Metric)=200°C
10 mm
14 mm
-60°C