Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Marca: Trajan Scientific and Medical™ 073273
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifica
SilTite Kit 1/32 in. Column for Agilent | |
1/32 in. column size | |
Installation for Capillary Columns |