missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35A3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553292
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: SSRSVLSPVV GTDAPDQHLE LKKPQELKEM ERLPLANEDK TMF Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 37%.
Uridine diphosphate-N-acetylglucosamine (UDP-GlcNAc) transporter in the Golgi apparatus. May supply UDP-GlcNAc as substrate for Golgi-resident glycosyltransferases that generate branching of diantennary oligosaccharides.Specifica
SLC35A3 | |
Polyclonal | |
Unconjugated | |
SLC35A3 | |
AMRS; golgi UDP-GlcNAc transporter; Slc35a3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3; solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member A3; solute carrier family 35 member 3; solute carrier family 35 member 3A; solute carrier family 35 member A3; UDP N-acetylglucosamine transporter; UDP-N-acetylglucosamine transporter | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2D2 | |
SLC35A3 | |
Recombinant protein corresponding to Human SLC35A3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only