Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix SM
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix SM fastens horizontal separating funnels of different sizes to platform.
Marca: Edmund Bühler GmbH™ 0051 484
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Fastens horizontal separating funnels of different sizes to platform
- For use with Combifix VKS or SM Universal Shakers
Order Info
Size
Specifica
Sliding Plate | |
Combifix SM | |
5 Clamping Strips H, additional base plate |