Learn More
3M™ Safety-Walk™ Slip-Resistant Fine Resilient Tape, Series 280
High friction, fine resilient textured material. Reverse side is coated with pressure sensitive adhesive protected by a removable liner. Slip resistant for wet, barefoot traffic areas.
Marca: 3M™ Safety-Walk™ 7000126122
Dettagli aggiuntivi : Peso : 0.71850kg
Specifica
18.3 x 0.051 m | |
60 ft. | |
White | |
Floor Tape | |
18.3 m |
2 in. | |
Acrylic | |
Wet, Barefoot Traffic Areas | |
0.051 m |