missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11156-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
SMARCD2 is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.Specifica
SMARCD2 | |
Polyclonal | |
Unconjugated | |
SMARCD2 | |
BAF60B, CRACD2, PRO2451, Rsc6p, SMARCD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6603, 83796, 83833 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92925, Q99JR8 | |
SMARCD2 | |
SMARCD2 Fusion Protein Ag1630 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |