missing translation for 'onlineSavingsMsg'
Learn More

SMCP Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA557156

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15775051

  • 514.00€ / 100µL
Spedizione stimata: 30-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: KHSKCCPAKG NQCCPPQQNQ CCQSKGNQCC PPKQNQCCQP KGSQCCPPKH NHCCQPKPPC CIQARCCGLE TKPEVSPLNM ESEPNSPQ Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 39%.

Involved in sperm motility. Its absence is associated with genetic background dependent male infertility. Infertility may be due to reduced sperm motility in the female reproductive tract and inability to penetrate the oocyte zona pellucida.
Specifica

Specifica

SMCP
Polyclonal
Unconjugated
SMCP
HSMCSGEN1; MCS; MCSP; mitochondrial capsule selenoprotein; SMCP; sperm mitochondria associated cysteine rich protein; sperm mitochondria-associated cysteine-rich protein; sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161
Rabbit
Antigen affinity chromatography
RUO
4184
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P49901
SMCP
Recombinant protein corresponding to Human SMCP
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only