Learn More
SOD1 Mouse anti-Human, Unconjugated, Clone: 2F10G1, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech 67480-1-IG-20UL
Codice prodotto. 16800136
  • 145.00€ / 20µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
The SOD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOD1. This antibody recognizes human, mouse, rat, pig antigen. The SOD1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifica
Specifica
SOD1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
2F10G1
Unconjugated
20 μL
6647
IgG2a