missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Calcium Exchanger 1/NCX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-93088-0.02ml
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sodium Calcium Exchanger 1/NCX1 Polyclonal antibody specifically detects Sodium Calcium Exchanger 1/NCX1 in Human samples. It is validated for Western Blot
Specifica
| Sodium Calcium Exchanger 1/NCX1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ37694, FLJ43417, Na(+)/Ca(2+)-exchange protein 1, sodium/calcium exchanger 1, solute carrier family 8 (sodium/calcium exchanger), member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-520 of human SLC8A1 (NP_066920.1). WVADRRLLFYKYVYKRYRAGKQRGMIIEHEGDRPSSKTEIEMDGKVVNSHVENFLDGALVLEVDERDQDDEEARREMARILKELKQKHPDKEIEQLIELANYQVLSQQQKSRAFYRIQATRLMTGAGNILKRHAADQARKAVSMHEVNTEVTENDPVSKIFFEQGTYQCLENCGTVALTIIRRGGDLTNTVFVDFRTEDGTANAGSDYEFTEGTVVFKPGDTQKEIRVGIIDDDIFEEDENFLVHLSNVKVSSEASEDGILEANHVSTLAC | |
| 0.02 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 6546 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto