Learn More
Sonic Hedgehog/Shh Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Marca:  Novus Biologicals NBP2-22139SS
 vedi altre versioni di questo prodotto
Codice prodotto. 18770614
  • 271.00€ / 25µL
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Sonic Hedgehog/Shh Polyclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Western Blot, Immunohistochemistry
Western Blot 1-2 ug/ml, Immunohistochemistry 1:200 - 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200 - 1:400
A synthetic peptide made to an N-terminal portion of the human SHH protein (between residues 1-75) [UniProt Q15465]
Human, Mouse
IgG
Polyclonal
HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
0.025 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni