Learn More
Sonic Hedgehog/Shh Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Marca: Novus Biologicals NBP2-22139
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Sonic Hedgehog/Shh Polyclonal specifically detects Sonic Hedgehog/Shh in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| A synthetic peptide made to an N-terminal portion of the human SHH protein (between residues 1-75) [UniProt Q15465] | |
| 0.1 mL |
Titolo del prodotto
For Research Use Only