Learn More
SOX2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11064-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is a rabbit polyclonal antibody raised against an internal region of human SOX2. a rare undifferentiated cell population that is intermingled with the Bergmann glia of the adult murine cerebellar cortex, expresses the stem cell markers Sox2
SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintece in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.Specifica
SOX2 | |
Polyclonal | |
Unconjugated | |
SOX2 | |
ANOP3, MCOPS3, SOX2, Transcription factor SOX 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20674, 378723, 499593, 6657 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P48431, P48432, Q6P0E1 | |
sox2 | |
SOX2 Fusion Protein Ag1530 | |
150 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |