missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG11A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16770-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody is specific to SPAG11.
Sperm binding proteins and their C-terminal peptides of the Sperm Associated Antigen 11 (SPAG11) family are found to play an important role in epididymal innate immunity in addition to their role in sperm maturation. SPAG11 peptides resulting due to cleavage at the furin-like proprotein convertase motif are reported in the epididymal fluid, ejaculate and on sperm and their antimicrobial activity demonstrated. SPAG11A belongs to this family and is also named as EP2, HE2. It has 4 isoforms produced by alternative splicing.Specifica
| SPAG11A | |
| Polyclonal | |
| Unconjugated | |
| SPAG11A | |
| EP2, HE2, Protein EP2, SPAG11A, Sperm antigen HE2, sperm associated antigen 11A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 653423 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PDA7 | |
| SPAG11A | |
| SPAG11A Fusion Protein Ag9846 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto