missing translation for 'onlineSavingsMsg'
Learn More

SPAG4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19721-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899194

  • 121.38€ / 20µL

Spedizione stimata: 04-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This antibody is specific to SPAG4.

The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development.
Specifica

Specifica

SPAG4
Polyclonal
Unconjugated
SPAG4
SPAG4, sperm associated antigen 4, SUN4
Rabbit
Antigen Affinity Chromatography
RUO
245865, 6676
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9JJF2, Q9NPE6
SPAG4
Peptide
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali